![Monoclonal Anti-Interferon-γ−Carboxyfluorescein antibody produced in mouse clone 25723.11, purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/374/370/0f604794-0c27-451a-bb16-d60dfe3d30b0/640/0f604794-0c27-451a-bb16-d60dfe3d30b0.jpg)
Monoclonal Anti-Interferon-γ−Carboxyfluorescein antibody produced in mouse
![Monoclonal Anti-Interferon-γ−Carboxyfluorescein antibody produced in mouse clone 25723.11, purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/374/370/0f604794-0c27-451a-bb16-d60dfe3d30b0/640/0f604794-0c27-451a-bb16-d60dfe3d30b0.jpg)
Synonym(s):
Monoclonal Anti-Interferon-γ
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing