
Monoclonal Anti-Hamster IgG−Peroxidase antibody produced in mouse

Synonym(s):
Monoclonal Anti-Hamster IgG
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing