Skip to Content
Merck
Methyl (S)-Boc-5-pyrrolidone-2-carboxylate 97%

Methyl (S)-Boc-5-pyrrolidone-2-carboxylate

Methyl (S)-Boc-5-pyrrolidone-2-carboxylate 97%
Synonym(s):
N-Boc-(S)-2-pyrrolidone-5-carboxylic acid methyl ester, Boc-L-pyroglutamic acid methyl ester, Methyl (S)-Boc-2-pyrrolidone-5-carboxylate, Methyl Boc-L-pyroglutamate
Empirical Formula (Hill Notation):
C11H17NO5
CAS Number:
Molecular Weight:
243.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing