Skip to Content
Merck
Methyl phenylsulfonylacetate 97%

Methyl phenylsulfonylacetate

Methyl phenylsulfonylacetate 97%
Synonym(s):
Methyl benzenesulfonylacetate
Linear Formula:
C6H5SO2CH2COOCH3
CAS Number:
Molecular Weight:
214.24

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing