Skip to Content
Merck
Methyl anthranilate ≥98%, FCC, FG

Methyl anthranilate

Methyl anthranilate ≥98%, FCC, FG
Synonym(s):
Methyl 2-aminobenzoate, Methyl anthranilate
Linear Formula:
2-(H2N)C6H4CO2CH3
CAS Number:
Molecular Weight:
151.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing