Skip to Content
Merck
Methoxycarbonylsulfenyl chloride 97%

Methoxycarbonylsulfenyl chloride

Methoxycarbonylsulfenyl chloride 97%
Synonym(s):
S-Chloro O-methyl thiocarbonate
Linear Formula:
CH3OCOSCl
CAS Number:
Molecular Weight:
126.56
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing