Skip to Content
Merck
Mesitaldehyde contains ~0.1% Hydroquinone as stabilizer, 98%

Mesitaldehyde

Mesitaldehyde contains ~0.1% Hydroquinone as stabilizer, 98%
Synonym(s):
2,4,6-Trimethylbenzaldehyde, 2-Formyl-1,3,5-trimethylbenzene, 2-Formylmesitylene
Linear Formula:
(CH3)3C6H2CHO
CAS Number:
Molecular Weight:
148.20

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing