Skip to Content
Merck
Mercury(II) perchlorate hydrate 99.998% trace metals basis

Mercury(II) perchlorate hydrate

Mercury(II) perchlorate hydrate 99.998% trace metals basis
Linear Formula:
Hg(ClO4)2·xH2O
CAS Number:
Molecular Weight:
399.49 (anhydrous basis)

Products

Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing