Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mercury dibromofluorescein disodium salt
Synonym(s):
2′,7′-Dibromo-5′-(hydroxymercurio)fluorescein disodium salt, Merbromin, Mercurochrome
Empirical Formula (Hill Notation):
C
20
H
8
Br
2
HgNa
2
O
6
CAS Number:
129-16-8
Molecular Weight:
750.65
EC Number:
204-933-6
Products
Product Category
dyes (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Physical Form
Special Grade
Practical (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
M7011
Mercury dibromofluorescein disodium salt,
practical grade, crystalline
Expand
View Pricing