Skip to Content
Merck
(−)-Menthyloxyacetic acid 98%

(−)-Menthyloxyacetic acid

(−)-Menthyloxyacetic acid 98%
Synonym(s):
(−)-Menthyl carboxymethyl ether
Empirical Formula (Hill Notation):
C12H22O3
CAS Number:
Molecular Weight:
214.30
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing