Skip to Content
Merck
Meclocycline sulfosalicylate salt

Meclocycline sulfosalicylate salt

Meclocycline sulfosalicylate salt
Empirical Formula (Hill Notation):
C22H21ClN2O8 · C7H6O6S
CAS Number:
Molecular Weight:
695.05
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing