Skip to Content
Merck
Malvidin-3-galactoside chloride phyproof® Reference Substance

Malvidin-3-galactoside chloride

Malvidin-3-galactoside chloride phyproof® Reference Substance
Synonym(s):
Arthanitin chloride, Primulin
Empirical Formula (Hill Notation):
C23H25ClO12
CAS Number:
Molecular Weight:
528.89

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2