Skip to Content
Merck
Magnesium phosphate tribasic octahydrate purum p.a., ≥97.0% (KT)

Magnesium phosphate tribasic octahydrate

Magnesium phosphate tribasic octahydrate purum p.a., ≥97.0% (KT)
Synonym(s):
Trimagnesium phosphate octahydrate
Linear Formula:
Mg3(PO4)2 · 8H2O
CAS Number:
Molecular Weight:
406.98
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing