
Lithium phenylacetylide solution

Synonym(s):
(Phenylethynyl)lithium
Linear Formula:
C6H5C≡CLi
CAS Number:
Molecular Weight:
108.07
Products
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing