Lithium isopropoxide solution
Synonym(s):
Lithium 2-propanolate
Linear Formula:
(CH3)2CHOLi
CAS Number:
Molecular Weight:
66.03
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing