Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium D-lactate
Synonym(s):
(
R
)-2-Hydroxypropionic acid lithium salt, D-Lactic acid lithium salt
Linear Formula:
CH
3
CH(OH)CO
2
Li
CAS Number:
27848-81-3
Molecular Weight:
96.01
EC Number:
248-693-0
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Purity
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
L1000
Lithium D-lactate,
95-100%
Expand
View Pricing