Lipoprotein, very low density from human plasma
Synonym(s):
Pre-β-lipoprotein, VLDL, Very low density lipoprotein
CAS Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing