Levetiracetam impurity B
Synonym(s):
Levetiracetam Crotonamide, (2Z)-2-(2-Oxopyrrolidin-1-yl)but-2-enamide
Empirical Formula (Hill Notation):
C8H12N2O2
CAS Number:
Molecular Weight:
168.19
Products
Formula Weight
Format
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing