Skip to Content
Merck
Lenalidomide ≥95%

Lenalidomide

Lenalidomide ≥95%
Synonym(s):
Lenalidomide
Empirical Formula (Hill Notation):
C13H13N3O3
CAS Number:
Molecular Weight:
259.26

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing