Skip to Content
Merck
KT182 ≥98% (HPLC)

KT182

KT182 ≥98% (HPLC)
Synonym(s):
[4-[3′-(Hydroxymethyl)[1,1′-biphenyl]-4-yl]-1H-1,2,3-triazol-1-yl](2-phenyl-1-piperidinyl)-methanone
Empirical Formula (Hill Notation):
C27H26N4O2
CAS Number:
Molecular Weight:
438.52

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing