Skip to Content
Merck
Kanamycin sulfate Streptomyces kanamyceticus powder, γ-irradiated

Kanamycin sulfate Streptomyces kanamyceticus

Kanamycin sulfate Streptomyces kanamyceticus powder, γ-irradiated
Synonym(s):
Kanamycin sulfate from Streptomyces kanamyceticus, Kanamycin A, Kanamycin sulfate salt
Empirical Formula (Hill Notation):
C18H36N4O11 · H2O4S
CAS Number:
Molecular Weight:
582.58
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Activity Spectrum
Microorganism Suitability
Mode of Action
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing