Skip to Content
Merck
Isopropyl nitrate purum, ≥98.0% (GC)

Isopropyl nitrate

Isopropyl nitrate purum, ≥98.0% (GC)
Linear Formula:
(CH3)2CHONO2
CAS Number:
Molecular Weight:
105.09
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing