Skip to Content
Merck
Isethionic acid sodium salt 98%

Isethionic acid sodium salt

Isethionic acid sodium salt 98%
Synonym(s):
2-Hydroxyethanesulfonic acid sodium salt
Linear Formula:
HOCH2CH2SO3Na
CAS Number:
Molecular Weight:
148.11
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing