Skip to Content
Merck
Iodo(triethyl phosphite)copper(I) 97%

Iodo(triethyl phosphite)copper(I)

Iodo(triethyl phosphite)copper(I) 97%
Synonym(s):
Copper(I) iodide-triethyl phosphite
Empirical Formula (Hill Notation):
C6H16CuIO3P
CAS Number:
Molecular Weight:
357.61

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing