Skip to Content
Merck
Indole-4-carboxaldehyde 97%

Indole-4-carboxaldehyde

Indole-4-carboxaldehyde 97%
Synonym(s):
4-Formylindole, NSC 337264
Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing