Skip to Content
Merck
Imidazo[1,2-a]pyridine-2-carboxylic acid technical grade, 85%

Imidazo[1,2-a]pyridine-2-carboxylic acid

Imidazo[1,2-a]pyridine-2-carboxylic acid technical grade, 85%
Empirical Formula (Hill Notation):
C8H6N2O2
CAS Number:
Molecular Weight:
162.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing