
ICI 204,448 hydrochloride

Synonym(s):
(±)-1-[2,3-(Dihydro-7-methyl-1H-inden-4-yl)oxy]-3-[(1-methylethyl)amino]-2-butanol hydrochloride, R,S-[3-[1-[[(3,4-Dichlorophenyl)acetyl]methylamino]-2-(1-pyrrolidinyl)ethyl] phenoxy]-acetic acid hydrochloride
Empirical Formula (Hill Notation):
C23H26Cl2N2O4 · HCl
CAS Number:
Molecular Weight:
501.83
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing