
(Hydroxymethyl)polystyrene

Synonym(s):
Benzyl alcohol polymer-bound, Polystyrene crosslinked with divinylbenzene, hydroxymethylated
CAS Number:
Products
Brand
Physical Form
Functional Group
Cleavage Condition
Linker
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing