Skip to Content
Merck
Hexamethylenetetramine tribromide 97%

Hexamethylenetetramine tribromide

Hexamethylenetetramine tribromide 97%
Synonym(s):
Hexamethylenetetramine hydrobromide perbromide, Hexamethylenetetramine, compound with hydrogen tribromide
Empirical Formula (Hill Notation):
C6H12N4 · HBr · Br2
CAS Number:
Molecular Weight:
380.91

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing