Skip to Content
Merck
Hexaethylene glycol monohexadecyl ether BioXtra, ≥99.0% (TLC)

Hexaethylene glycol monohexadecyl ether

Hexaethylene glycol monohexadecyl ether BioXtra, ≥99.0% (TLC)
Synonym(s):
C16E6, Hexadecylhexaglycol
Linear Formula:
CH3(CH2)15(OCH2CH2)6OH
CAS Number:
Molecular Weight:
506.76

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing