Skip to Content
Merck
Heptadecafluorooctanesulfonic acid tetraethylammonium salt 98%

Heptadecafluorooctanesulfonic acid tetraethylammonium salt

Heptadecafluorooctanesulfonic acid tetraethylammonium salt 98%
Synonym(s):
Perfluorooctanesulfonic acid tetraethylammonium salt
Linear Formula:
CF3(CF2)6CF2SO3N(C2H5)4
CAS Number:
Molecular Weight:
629.37
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing