Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glycerol phosphate calcium salt hydrate
Empirical Formula (Hill Notation):
C
3
H
7
CaO
6
P · xH
2
O
CAS Number:
1336-00-1
Molecular Weight:
210.14 (anhydrous basis)
EC Number:
215-643-4
Products
Product Category
building blocks (1)
carbohydrates (1)
glycerols (1)
lipids (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Purum (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
50043
Glycerol phosphate calcium salt hydrate,
mixture of isomers of α- and β-form, ≥96% (calc. based on dry substance, T)
Expand
View Pricing
50057
Glycerol phosphate calcium salt hydrate,
tested according to Ph. Eur.
Expand
View Pricing