Fmoc-Tic-OH
Synonym(s):
(S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Fmoc-Tic-OH
Empirical Formula (Hill Notation):
C25H21NO4
CAS Number:
Molecular Weight:
399.44
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing