Skip to Content
Merck
Fmoc-D-Phe(4-F)-OH ≥98.0%

Fmoc-D-Phe(4-F)-OH

Fmoc-D-Phe(4-F)-OH ≥98.0%
Synonym(s):
Fmoc-4-fluoro-D-phenylalanine
Empirical Formula (Hill Notation):
C24H20FNO4
CAS Number:
Molecular Weight:
405.42

Products

Brand
Formula Weight
Purity
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing