
Fenclorim

Synonym(s):
4,6-Dichloro-2-phenylpyrimidine
Empirical Formula (Hill Notation):
C10H6Cl2N2
CAS Number:
Molecular Weight:
225.07
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing