Skip to Content
Merck
Fenchlorazol-ethyl PESTANAL®, analytical standard

Fenchlorazol-ethyl

Fenchlorazol-ethyl PESTANAL®, analytical standard
Synonym(s):
Ethyl 1-(2,4-dichlorophenyl)-5-(trichloromethyl)-1H-1,2,4-triazole-3-carboxylate
Empirical Formula (Hill Notation):
C12H8Cl5N3O2
CAS Number:
Molecular Weight:
403.48
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing