Skip to Content
Merck
Ethyl trimethylsilyl malonate 97%

Ethyl trimethylsilyl malonate

Ethyl trimethylsilyl malonate 97%
Linear Formula:
H3CCH2OCOCH2COOSi(CH3)3
CAS Number:
Molecular Weight:
204.30
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing