Skip to Content
Merck
Ethyl homovanillate 97%

Ethyl homovanillate

Ethyl homovanillate 97%
Synonym(s):
Ethyl 4-hydroxy-3-methoxyphenylacetate
Linear Formula:
HOC6H3(OCH3)CH2CO2C2H5
CAS Number:
Molecular Weight:
210.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing