Ethyl β-carboline-3-carboxylate
Synonym(s):
β-CCE, Ethyl 9H-pyrido[3,4-b]indole-3-carboxylate
Empirical Formula (Hill Notation):
C14H12N2O2
CAS Number:
Molecular Weight:
240.26
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing