Skip to Content
Merck
Ethyl 1,3-dithiolane-2-carboxylate ≥98.0%

Ethyl 1,3-dithiolane-2-carboxylate

Ethyl 1,3-dithiolane-2-carboxylate ≥98.0%
Synonym(s):
Glyoxylic acid ethyl ester ethylene mercaptal
Empirical Formula (Hill Notation):
C6H10O2S2
CAS Number:
Molecular Weight:
178.27
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing