
Ethychlozate

Synonym(s):
Ethyl 5-chloro-1H-indazole-3-acetate
Empirical Formula (Hill Notation):
C11H11ClN2O2
CAS Number:
Molecular Weight:
238.67
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing