Skip to Content
Merck
(Ethoxycarbonylmethyl)triphenylphosphonium chloride technical, ≥90% (AT)

(Ethoxycarbonylmethyl)triphenylphosphonium chloride

(Ethoxycarbonylmethyl)triphenylphosphonium chloride technical, ≥90% (AT)
Synonym(s):
(2-Ethoxy-2-oxoethyl)triphenylphosphonium chloride
Linear Formula:
CH3CH2OCOCH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
384.84
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing