Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S. set
Products
Product Category
pipette tips (6)
Manufacturer
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z640298
Eppendorf
®
epT.I.P.S. set,
volume range 0.1-10 μL
Expand
View Pricing
Z640352
Eppendorf
®
epT.I.P.S. set,
volume range 50-1000 μL
Expand
View Pricing
Z640336
Eppendorf
®
epT.I.P.S. set,
volume range 2-200 μL
Expand
View Pricing
Z640344
Eppendorf
®
epT.I.P.S. set,
volume range 20-300 μL
Expand
View Pricing
Z640360
Eppendorf
®
epT.I.P.S. set,
volume range 50-1250 μL
Expand
View Pricing
Z640328
Eppendorf
®
epT.I.P.S. set,
volume range 0.5-20 μL
Expand
View Pricing