
DMT-2′O-TBDMS-rG(tac) Phosphoramidite

Synonym(s):
DMT-2′-O-TBDMS-rG(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-guanosine,3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C58H76N7O10PSi
CAS Number:
Molecular Weight:
1090.32
Products
Product Category
Brand
Formula Weight
Product Line
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing