
(±)-8a-Methyl-3,4,8,8a-tetrahydro-1,6(2H,7H)-naphthalenedione

Synonym(s):
(±)-9-Methyl-5(10)-octaline-1,6-dione, Miescher-Wieland ketone, Wieland-Miescher ketone
Empirical Formula (Hill Notation):
C11H14O2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing