
(±)-6-Methyl-5,6,7,8-tetrahydropterine dihydrochloride

Synonym(s):
6-MPH4, DL-2-Amino-4-hydroxy-6-methyl-5,6,7,8-tetrahydropteridine dihydrochloride
Empirical Formula (Hill Notation):
C7H11N5O · 2HCl
CAS Number:
Molecular Weight:
254.12
Products
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing