Skip to Content
Merck
Discovery® Zr-PS HPLC Column 3 μm particle size, L × I.D. 15 cm × 4.6 mm

Discovery® Zr-PS HPLC Column

Discovery® Zr-PS HPLC Column 3 μm particle size, L × I.D. 15 cm × 4.6 mm

Products

Product Category
Brand
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1