Skip to Content
Merck
Dimethyl trans-cyclohexane-1,4-dicarboxylate 99%

Dimethyl trans-cyclohexane-1,4-dicarboxylate

Dimethyl trans-cyclohexane-1,4-dicarboxylate 99%
Linear Formula:
C6H10(CO2CH3)2
CAS Number:
Molecular Weight:
200.23

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing