Skip to Content
Merck
Dimethyl (4R,5R)-1,3,2-dioxathiolane-4,5-dicarboxylate 2,2-dioxide 97%

Dimethyl (4R,5R)-1,3,2-dioxathiolane-4,5-dicarboxylate 2,2-dioxide

Dimethyl (4R,5R)-1,3,2-dioxathiolane-4,5-dicarboxylate 2,2-dioxide 97%
Synonym(s):
(−)-Dimethyl 2,3-O-sulfonyl-L-tartrate, Dimethyl L-tartrate cycl-sulfate
Empirical Formula (Hill Notation):
C6H8O8S
CAS Number:
Molecular Weight:
240.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing