
Diloxanide furoate

Synonym(s):
2,2-Dichloro-N-(4-hydroxyphenyl)-N-methylacetamide 2-furoic acid ester
Empirical Formula (Hill Notation):
C14H11Cl2NO4
CAS Number:
Molecular Weight:
328.15
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing